Sodium benzenesulfinate ≥98%
Supplier: Matrix Scientific
Synonyms:
Benzenesulphinic acid sodium salt
, Benzenesulfinic acid sodium salt
, Benzenesulphinic acid, sodium salt
, Benzenesulfinic acid, sodium salt
004310-1KG
101787-084EA
223.08
USD
101787-084
Sodium benzenesulfinate ≥98%
Sodium benzenesulphinate
Hazard code(s): Irritant
Formula:
C₆H₅NaO₂S MW: 164.16 g/mol Melting Pt: >300 °C |
MDL Number:
MFCD00013135 CAS Number: 873-55-2 EINECS: 212-842-8 |
Specification Test Results
Melting point |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...