5-Methoxybenzofuran for synthesis

Supplier: Matrix Scientific

003017-1G 003017-5G
101785-086EA 129.23 USD
101785-086 101785-088
5-Methoxybenzofuran for synthesis
5-Methoxybenzofuran

Hazard code(s): Irritant

Formula: C₉H₈O₂
Boiling Pt: 123 °C (20mm)
Melting Pt: 31…33 °C
MDL Number: MFCD00079770
CAS Number: 13391-28-1

Order Now

Specification Test Results

Boiling point 123°C/20mm
Melting point 31-33°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR