5-Nitroisatoic anhydride, tech.
Supplier: Matrix Scientific
Synonyms:
6-Nitro-2H-3,1-benzoxazine-2,4(1H)-dione
002993-5G
002993-25G
101785-024EA
147.69
USD
101785-024
101785-026
5-Nitroisatoic anhydride, tech.
5-Nitroisatoic anhydride
Hazard code(s): Irritant
Formula:
C₈H₄N₂O₅ MW: 208.13 g/mol Melting Pt: 265…266 °C (dec.) |
MDL Number:
MFCD00023891 CAS Number: 20829-97-4 |
Specification Test Results
Melting point | 265-266°C(d |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...