(3-Fluoro-4-hydroxyphenyl)acetic acid ≥98%

Supplier: Matrix Scientific

002538-1G 002538-5G 002538-25G
101784-096EA 38.46 USD
101784-096 101784-098 101784-100
(3-Fluoro-4-hydroxyphenyl)acetic acid ≥98%
(3-Fluoro-4-hydroxyphenyl)acetic acid

Hazard code(s): Irritant

Formula: C₈H₇FO₃
Melting Pt: 132…134 °C
MDL Number: MFCD00004348
CAS Number: 458-09-3
EINECS: 207-275-8

Order Now

Specification Test Results

Melting point 132-134°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR