4-Nitro-2-(trifluoromethyl)aniline ≥98% for synthesis
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
2-Amino-5-nitro-α,α,α-trifluortoluene, 2-Amino-5-nitro-α,α,α-trifluorotoluene, 4-Nitro-2-trifluoromethylaniline, 2-Amino-5-nitrobenzotrifluoride, 4-Nitro-α,α,α-trifluoro-o-toluidine
001845-25G
001845-100G
101782-712EA
33.85
USD
101782-712
101782-714
4-Nitro-2-(trifluoromethyl)aniline ≥98% for synthesis
4-Nitro-2-(trifluoromethyl)aniline
Hazard code(s): Irritant
Formula:
C₇H₅F₃N₂O₂ Melting Pt: 90…92 °C Density: 1.754 g/cm³ (–123.15 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007365 CAS Number: 121-01-7 EINECS: 204-443-2 UN: 2811 ADR: 6.1,III |
Specification Test Results
Melting point | 90-92°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...