2-Amino-5-iodopyridine ≥98% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: 5-Iodopyridin-2-amine, 5-Iodo-2-pyridylamine

001779-5G 001779-25G
101782-560EA 15.38 USD
101782-560 101782-562
2-Amino-5-iodopyridine ≥98% for synthesis
2-Amino-5-iodopyridine

Hazard code(s): Irritant

Formula: C₅H₅IN₂
Boiling Pt: 271 °C (1013 hPa)
Melting Pt: 128…130 °C
MDL Number: MFCD00160312
CAS Number: 20511-12-0

Order Now

Specification Test Results

Melting point 128-130°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR