6-Fluoroanthranilic acid ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 2-Amino-6-fluorobenzoic acid

001696-5G 001696-100G 001696-25G
101782-306EA 23.08 USD
101782-306 101782-308 101782-310
6-Fluoroanthranilic acid ≥97%
6-Fluoroanthranilic acid

Hazard code(s): Irritant

Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 165…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00067781
CAS Number: 434-76-4

Order Now

Specification Test Results

Melting point 165-167°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR