3-Amino-4-bromo-1H-pyrazole ≥97% for synthesis
Supplier: Matrix Scientific
Synonyms:
4-Bromo-1H-pyrazol-5-amine, 4-Bromo-1H-pyrazol-3-amine, 3-Amino-4-bromopyrazole
001595-1G
001595-5G
001595-25G
101782-062EA
29.23
USD
101782-062
101782-064
101782-066
3-Amino-4-bromo-1H-pyrazole ≥97% for synthesis
3-Amino-4-bromo-1H-pyrazole
Hazard code(s): Irritant
Formula:
C₃H₄BrN₃ Melting Pt: 133…135 °C |
MDL Number:
MFCD00082728 CAS Number: 16461-94-2 |
Specification Test Results
Melting point | 133-135°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...