2-Amino-3-bromo-5-methylpyridine ≥97% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: 3-Bromo-5-methylpyridin-2-amine, 2-Amino-3-bromo-5-picoline, 3-Bromo-5-methyl-2-pyridylamine

001587-25G 001587-100G
101782-030EA 44.62 USD
101782-030 101782-032
2-Amino-3-bromo-5-methylpyridine ≥97% for synthesis
2-Amino-3-bromo-5-methylpyridine

Hazard code(s): Irritant

Formula: C₆H₇BrN₂
Melting Pt: 74…75 °C
MDL Number: MFCD00068231
CAS Number: 17282-00-7

Order Now

Specification Test Results

Melting point 74-75°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR