2,3,4,5,6-Pentafluorobenzyl alcohol ≥99%
Supplier: Matrix Scientific
|
Warning
|
001553-5G
001553-25G
001553-100G
101781-954EA
21.54
USD
101781-954
101781-956
101781-958
2,3,4,5,6-Pentafluorobenzyl alcohol ≥99%
2,3,4,5,6-Pentafluorobenzyl alcohol
Hazard code(s): Irritant
Formula:
C₇H₃F₅O Boiling Pt: 114…115 °C (60mm) Melting Pt: 37…38 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004602 CAS Number: 440-60-8 EINECS: 207-126-7 |
Specification Test Results
Boiling point | 114-115°C/6 |
Melting point | 37-38°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...