1-Bromo-2,4,6-trifluorobenzene ≥99%
Supplier: Matrix Scientific
|
Warning
|
001227-25G
001227-100G
101781-208EA
180
USD
101781-208
101781-210
1-Bromo-2,4,6-trifluorobenzene ≥99%
1-Bromo-2,4,6-trifluorobenzene
Hazard code(s): Irritant
Formula:
C₆H₂BrF₃ Boiling Pt: 140.5 °C Melting Pt: 3.5 °C Density: 1.790 g/cm³ |
MDL Number:
MFCD00000334 CAS Number: 2367-76-2 EINECS: 219-125-9 UN: 1993 ADR: 3,III |
Specification Test Results
Boiling point | 140.5°C |
Melting point | 3.5°C |
Density | 1.79 |
ND20 | 1.485 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...