Potassium thiocyanate 10% (w/v) in aqueous solution

Supplier: Ricca Chemical

Synonyms: Potassium rhodanide

R6555000-500A R6555000-4A R6555000-1A
RC6555-32EA 149.64 USD
RC6555-32 RC6555-16 RC6555-1
Potassium thiocyanate 10% (w/v) in aqueous solution
Potassium thiocyanate
Formula: KSCN
MW: 97.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011413
CAS Number: 333-20-0
EINECS: 206-370-1

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Refractive Index Confirmed concentration

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR