Sodium thiocyanate 0.100 N (N/10) in aqueous solution
Supplier: Ricca Chemical
|
Warning
|
R7860000-10F
R7860000-20F
R7860000-4A
R7860000-1A
RC786025EA
203
USD
RC786025
RC7860-5
RC7860-1
RC7860-32
Sodium thiocyanate 0.100 N (N/10) in aqueous solution
Sodium thiocyanate
Formula:
NaSCN Storage Temperature: Ambient |
MDL Number:
MFCD00011123 CAS Number: 540-72-7 EINECS: 231-867-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...