Sodium acetate 0.200 M (M/5) in aqueous solution analytical reagent

Supplier: Ricca Chemical

Synonyms: Acetic acid sodium salt

R7129000-500A R7129000-1A R7129000-4A
RC7129-16EA 76 USD
RC7129-16 RC7129-32 RC7129-1
Sodium acetate 0.200 M (M/5) in aqueous solution analytical reagent
Sodium acetate
Formula: H₃CCOONa
MW: 82.03 g/mol
Boiling Pt: 881.4
Melting Pt: 324
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
EINECS: 204-823-8
Merck Index: 12,08711

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Certified Concentration 0.200 ± 0.001 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR