2-Propanol 50% (v/v) in aqueous solution (1 + 1) analytical reagent

Supplier: Ricca Chemical
Danger

Synonyms: Isopropyl rubbing alcohol, sec-Propanol, Isopropanol, 2-Hydroxypropane, Isopropyl Rubbing Alcohol, 2-Hydroxy propane, IPA, sec-Propyl Alcohol, Dimethyl carbinol, Propane, 2-hydroxy-, Isopropyl alcohol, Propan-2-ol

R4207500-4A R4207500-20E1 R4207500-20F
RC4207.5-1EA 168 USD
RC4207.5-1 RC4207.5-5HP RC4207.5-5
2-Propanol 50% (v/v) in aqueous solution (1 + 1) analytical reagent
2-Propanol
Formula: (CH₃)₂CHOH
Density: 0.8…0.92 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1987
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, IPA odor
Specific Gravity 50 ± 2 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR