Phenolphthalein 0.1% (w/v) in 80% alcohol (v/v) solution R
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
R5598000-500A
R5598000-1A
RC5598-16EA
61
USD
RC5598-16
RC5598-32
Phenolphthalein 0.1% (w/v) in 80% alcohol (v/v) solution R
Phenolphthalein
Formula:
C₂₀H₁₄O₄ MW: 318.33 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00005913 CAS Number: 77-09-8 EINECS: 201-004-7 ADR: 3,II |
Specification Test Results
Appearance | Clear, very light pink, alcohol odor |
Suitability for Use | Colorless (pH 8.0) - Pink (pH 10.0) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...