Orthoboric acid 2% (w/v) in aqueous solution for ammonia and Kjeldahl nitrogen analysis, with mixed indicator
Supplier: Ricca Chemical
Synonyms:
Boron trihydroxide, Orthoboric acid, Boron hydroxide, Boric acid, Boracic acid
R1064000-4A
R1064000-10F
R1064000-1A
RC1064-1EA
259
USD
RC1064-1
RC1064-2.5
RC1064-32
Orthoboric acid 2% (w/v) in aqueous solution for ammonia and Kjeldahl nitrogen analysis, with mixed indicator
ortho-Boric acid
Formula:
B(OH)₃ MW: 61.83 g/mol Boiling Pt: 185 Melting Pt: 160 Storage Temperature: Ambient |
MDL Number:
MFCD00011337 CAS Number: 10043-35-3 EINECS: 233-139-2 Merck Index: 13,01326 |
|
Specification Test Results
Absorbance (traceable to NIST SRM 930) | Conforms to control |
Appearance | Clear, dark blue, odorless |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...