Hydrochloric acid 0.200 N (N/5) analytical reagent

Supplier: Ricca Chemical

Synonyms: Hydrochloric acid solution, Muriatic acid

R3620000-4A R3620000-1A R3620000-20F
RC36201EA 112 USD
RC36201 RC362032 RC36205
Hydrochloric acid 0.200 N (N/5) analytical reagent
Hydrochloric acid
Formula: HCl
MW: 36.46
Density: 1.004 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 0.2000 ± 0.0001 N at 20.0 °C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR