Nitric acid 0.500 N (N/2)

Supplier: Ricca Chemical

Synonyms: Nitric acid solution

R5425000-20F
RC54255EA 758.64 USD
RC54255
Nitric acid 0.500 N (N/2)
Nitric acid
Formula: HNO₃
MW: 63.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 0.5000 ± 0.0005 N at 20.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR