Nitric acid 0.0100 N (N/100)

Supplier: Ricca Chemical

Synonyms: Nitric acid solution

R5380000-500A R5380000-1A
RC5380-32EA 61.79 USD
RC5380-32 RC5380-16
Nitric acid 0.0100 N (N/100)
Nitric acid
Formula: HNO₃
MW: 63.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 0.01000 ± 0.00002 N at 20.0°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR