Potassium chloride 3 M saturated with silver chloride aq sol Electrodes_filling
Supplier: Ricca Chemical
Synonyms:
Potassiumchloride
R5918000-500A
R5918000-1A
RC5918-16EA
71
USD
RC5918-16
RC5918-32
Potassium chloride 3 M saturated with silver chloride aq sol Electrodes_filling
Potassium chloride
Formula:
KCl MW: 74.55 g/mol Boiling Pt: 1420 Melting Pt: 773 Density: 1.13 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011360 CAS Number: 7447-40-7 EINECS: 231-211-8 Merck Index: 14,07621 |
Specification Test Results
Appearance | Clear, colorless, odorless |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...