Sodium hydroxide 40% (w/w) in aqueous solution analytical reagent

Supplier: Ricca Chemical
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

R7288000-20F R7288000-1A R7288000-10F R7288000-500A
RC7288-5EA 418 USD
RC7288-5 RC7288-32 RC7288-2.5 RCR7288000500A
Sodium hydroxide 40% (w/w) in aqueous solution analytical reagent
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Order Now

Specification Test Results

Appearance Colorless liquid
Assay at 20 °C (traceable to NIST SRM 723) 39.8 - 40.2 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR