Potassium permanganate 0.0500 N (N/20) in aqueous solution

Supplier: Ricca Chemical

Synonyms: Permanganic acid potassium salt

R6390000-1C R6390000-4C
RC6390-1EA 153.65 USD
RC6390-1 RC6390-32
Potassium permanganate 0.0500 N (N/20) in aqueous solution
Potassium permanganate
Formula: KMnO₄
MW: 158.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear, purple, odorless
Assay at 20 °C (traceable to NIST SRM 40) 0.05000 ± 0.00005 N at 20.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR