Potassium hydroxide 0.100 N (N/10) in denatured alcohol
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Potash caustic
, Potassium hydrate
, Caustic potash
R6264500-1A
R6264500-4A
RC6264.5-1EA
303.07
USD
RC6264.5-1
RC6264.5-32
Potassium hydroxide 0.100 N (N/10) in denatured alcohol
Potassium hydroxide
Formula:
KOH MW: 56.11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003553 CAS Number: 1310-58-3 EINECS: 215-181-3 UN: 2924 ADR: 3,II |
Specification Test Results
Appearance | Clear, colorless, alcohol odor |
Assay at 20 °C (traceable to NIST SRM 723) | 0.1000 ± 0.0005 N at 20.0 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...