Oxalic acid 5% w/v aqueous solution zA
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Ethanedioic acid
R5465500-500A
R5465500-1A
RC5465.5-16EA
64
USD
RC5465.5-16
RC5465.5-32
Oxalic acid 5% w/v aqueous solution zA
Oxalic acid
Formula:
C₂H₂O₄ MW: 90.04 g/mol Boiling Pt: 365 Storage Temperature: Ambient |
MDL Number:
MFCD00002573 CAS Number: 144-62-7 EINECS: 205-634-3 UN: 3265 ADR: 8 III Merck Index: 13,06980 |
Specification Test Results
Appearance | Clear, colorless, odorless |
Refractive Index | 5.0 ± 0.5 % (w/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...