Barium chloride 20% (w/v) in aqueous solution
Supplier: Ricca Chemical
|
Warning
|
R0855000-1A
R0855000-20F
R0855000-4A
R0855000-500A
RC855-32EA
97
USD
RC855-32
RC855-5
RC855-1
RC855-16
Barium chloride 20% (w/v) in aqueous solution
Barium chloride
Formula:
BaCl₂ Boiling Pt: ∼100 °C (1013 hPa) Storage Temperature: Ambient |
MDL Number:
MFCD00003445 CAS Number: 10361-37-2 EINECS: 233-788-1 UN: 3287 ADR: 6.1,III |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay | 20.0 ± 0.5 % w/v |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...