Protein Detector™ Phosphatase ELISA Kits, Lgc Clinical Diagnostics, Inc
Supplier: LGC Clinical Diagnostics
Protein Detector™
5110-0012
95059-364EA 581.15 USD
95059-364
Protein Detector™ Phosphatase ELISA Kits, Lgc Clinical Diagnostics, Inc
Assays ELISAs
AP ELISA Kits offer a convenient starting point for the development of microwell ELISA protocols for antigen and antibody detection
Kit contains Coating Solution Concentrate; BSA Diluent/Blocking Solution Concentrate; Wash Solution Concentrate; 50% Glycerol Solution; AP labeled Goat Anti-Human IgG (H+L); AP labeled Goat Anti-Rabbit IgG (H+L); BluePhos Phosphatase Substrate System; and APstop Solution Concentrate.
Highly sensitive reagents are optimized for antigen detection in picogram quantities. The wash and block solutions ensure low background. This kit can be used for direct and indirect detection.
These kits provide pretested components and suggested protocols to facilitate the development of ELISA procedures.