Vacule® Vial, Wheaton, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
651954 651907
16300-200CS 294.07 USD
16300-200 26675-909 26675-920
Vacule® Vial, Wheaton, DWK Life Sciences
Vials
The Wheaton Vacule® Vial features heavy wall construction for lyophilization and freeze-drying applications.
Vials are manufactured from Wheaton 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E438 Type I, Class A requirements.
Vacule® Vials also feature a controlled ID for accurate stopper fit and screw threads that eliminate the use of an aluminum seal.