DuPont™ Tychem® 2000 Bib Aprons
Supplier: DuPont
Tychem®
QC273BYL00010000
11000-498CS 366.04 USD
11000-498
DuPont™ Tychem® 2000 Bib Aprons
Disposable Garments
A lightweight, and durable fabric, DuPont™ Tychem® 2000 utilizes the strength of DuPont™ Tyvek® fabric and a polyethylene coating.
  • Neck loop
  • Waist ties
  • Bound seams
Tychem® 2000 fabric provides at least 30 minutes of protection against 42 chemical challenges. Tychem® 2000 is used for light splash protection in a variety of industrial environments, including petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing. Bib aprons feature neck loop and waist ties with bound seams.
Certifications: Thickness meets ASTM D1777.