Zinc Nitrate 6-Hydrate

Supplier: ALDON CORP SE

ZZ0080-500G ZZ0080-500GR ZZ0080-CAPS ZZ0080-PK/4R ZZ0092-CONC ZZ0091-100ML ZZ0094-500ML ZZ0092-500ML
470303-120EA 39.2 USD
470303-120 470303-124 470303-118 470303-126 470303-128 470303-132 470225-668 470328-166
Zinc Nitrate 6-Hydrate
Chemicals for Science Education

CAS Number: 10196-18-6
Formula: ZnNO32 6H2O
Density: 2.065 g/mL
Boiling Point: 105-130°C
Freezing Point: 36.4°C
Solubility: Water and Alcohol
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This product is a toxic oxidizer.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR