Zea, Branch Root Slide
Supplier: Avantor
WARD470181-290
470181-290EA 8.45 USD
470181-290
Zea, Branch Root Slide
Slides Prepared Slides
Root Development
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Corn. Shows origin of the lateral root in vascular cylinder.