Wandering Jew
Supplier: CHASE GREENHOUSES, INC.
867300
470180-026EA 7.35 USD
470180-026
Wandering Jew
Specimens Live Specimens Plants - Living Material
Tradescantia virginiana or similar. Monocot. This hardy, easy-to-grow ornamental plant can be used for studying meiotic division in mother cells of pollen. Cytoplasmic flows can be seen in cell hairs on staminate filaments. It is native to tropical and subtropical areas of the Americas and is supplied in a pot with care instructions. Size: 6".