The Reflex Arc Microslide
Supplier: AMERICAN EDUCATIONAL PROD
T-70
470107-924PK 10.3 USD
470107-924
The Reflex Arc Microslide
Slides Prepared Slides
Illustrates nervous tissues and reflexive tissues
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects it on both sides
  • Set of 15 includes detailed lesson plan on the topic under study
Reflex acts are inborn; not learned by imitation or study. These acts include the eye blinking automatically when anything moves towards it, the flow of saliva produced when food is placed in the mouth and a sneeze caused by an irritating particle in the nasal passage. This set traces a reflex arc through the nervous system.
Introduce junior and senior high school students to the world of microscopy with inexpensive, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides. A detailed text folder is included. Viewers are available separately.
Contents:
Pacinian corpuscle - cs/st (90X)
Pacinian corpuscle - cs/st (360X)
Spinal ganglion and nerve - cs/st (80X)
Spinal cord and nerve - cs/st (6X)
Spinal cord - cs/silver stained (10X)
Gray matter - cs/st (200X)
Motor neuron - cs/st (400X)
Motor end plates (on striated muscle) - cs/st (300X)