Taenia pisiformis, Mature/Gravid Proglottids Slide
Supplier: Avantor
WARD470182-112
470182-112EA 19.69 USD
470182-112
Taenia pisiformis, Mature/Gravid Proglottids Slide
Slides Prepared Slides
Endoparasitic Tapeworm
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Composite of mature and gravid proglottids of Taenia pisiformis. Mature proglottid shows uterus, ovary, testes, sperm duct, and genital pore. Gravid proglottid shows eggs.