Ward's® Student Bacterial Culture Kit
Supplier: VWR
WARD470030-200
470030-200EA 38.79 USD
470030-200
Ward's® Student Bacterial Culture Kit
Specimens Live Specimens Bacteria - Living Material
Everything you need to safetly collect and culture bacteria and fungi from your environment
  • Students collect and culture microorganisms from their own environment
  • Bunsen burner or other sterilizer not required
  • Comes with prepared media- just melt and pour
The equipment included in our convenient kit is ready to use, making bacterial culturing easy. Best of all, a Bunsen burner, used in standard inoculating procedures, is not required.
Kit includes:
Two bottles of nutrient agar
20 sterile mini-Petri dishes
20 sterile applicator swabs
autoclavable disposable bag
instructions
Ordering information: This item will ship immediately upon order, unless you specify otherwise.