Strontium Chloride 6-Hydrate

Supplier: ALDON CORP SE

SS0996-100G SS0996-500G SS0996-100GR SS0996-500GR SS0991-500ML
470302-780EA 15.1 USD
470302-780 470302-782 470302-784 470302-786 470302-788
Strontium Chloride 6-Hydrate
Chemicals for Science Education

CAS Number: 10025-70-4
Formula Weight: 266.62
Formula: SrCl2·6H2O
Density: 2.986 g/mL
Freezing Point: 570°C
Solubility: Water and Alcohol
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR