Ward's® Live Streptomyces violaceus Culture
Supplier: VWR
WARD470179-190
470179-190EA 12.95 USD
470179-190
Ward's® Live Streptomyces violaceus Culture
Specimens Live Specimens Bacteria - Living Material
Produces a violet color that diffuses through agar growth medium. Anti–bacterial, anti–fungal, anti–viral activities. Branching vegetative mycelium. 
  • Biosafety Level: 1
  • Motility: Negative
  • Morphology: Filamentous rods
  • Gram Reaction: Positive
  • Respiration: Aerobic
  • Chromogenesis: Yes (violet)
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
Ordering information: Microbial and fungal cultures are restricted and can only be ordered by educational professionals.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout
Caution: While this organism is non-pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.