Specific Gravity Balls Set
Supplier: TONAWANDA MANUFACTURING
4734601 47346-00 47346-00
470016-780KT 24.8 USD
470016-780 470014-692 470200-698
Specific Gravity Balls Set
Educational Materials Physics Educational Materials Mechanics Learning Activities
For your Laboratory Needs!
  • Choose from two different sets of specific gravity balls
  • Size of each ball: 3/4" dia.
  • Set of four
The Plastic Set contains one each of acrylic (Plexiglas®), acetal (Delrin®), nylon and polypropylene with a specific gravity of 1.19, 1.42, 1.16 and 0.91 respectively.
The Various Set contains one each of steel, polypropylene, HDPE and acrylic with a specific gravity of 7.7, 0.91, 0.954 and 1.19 respectively.