Somso® White Mould
Supplier: MARCUS SOMMER
Somso®
BOS 14/1
470029-840EA 348.19 USD
470029-840
Somso® White Mould
Models Human Biology Models
Somso® premium quality models.
  • Five year warranty
Mucor mucedo, enlarged approx. 250 times, in SOMSO-Plast®, according to Prof. Dr. W. Weber. The model shows sexual and asexual reproduction. Separable into three parts. Mounted on a green board with explanatory note.
Ordering information: Items on this page are not stocked and may have an extended time for delivery.