Sodium Alginate

Supplier: ALDON CORP SE

AA0099-25G AA0098-100ML
470302-432EA 20.65 USD
470302-432 470233-592
Sodium Alginate
Chemicals for Science Education

CAS Number: 9005-38-3
Formula Weight: Mixture
Formula: Mixture
Solubility: Hot Water
Synonyms: Alginic Acid
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR