Ward's® Silicate Mineral Specimens Card Set
Supplier: MERCURY PRINT PRODUCTIONS SE
362500
470023-888KT 6.49 USD
470023-888
Ward's® Silicate Mineral Specimens Card Set
Educational Materials Earth Science Educational Materials Earth Science Flashcards
There’s no more convenient way of learning essential properties of minerals than with this informative, decorative set of cards exclusively from WARD’S.
  • Size: 3½×2½"
Vivid photos and important information on a variety of mineral types are all contained on these trading-card-size reference cards, making them useful as flash cards to help students learn or review key properties and features, or as informational supplements to specimen collections. The mineral specimen cards list the mineral group, chemical composition, crystallography, physical properties, composition, occurrence, and uses.
Mineral groups: nesosilicates, sorosilicates, cyclosilicates, inosilicates, phyllosilicates, and tectosilicates.