Rapid Radish Seeds
Supplier: Avantor
WARD470003-928 WARD470003-930
470003-930PK 44.99 USD
470003-930 470003-928
Rapid Radish Seeds
Cell Culture Supplements and Additives
Rapid Radish seeds are selectively bred to germinate and grow quickly, giving you fast, accurate results.
The large seeds are easy to handle and also emerge easily from the soil, with a high germination rate. In addition, the plants, which remain hardy throughout their life cycle, feature large flowers that allow for the study of various structures.This item is NOT irradiated.