Preserved Rabbits
Supplier: Ward's Science
140.1057.1 140.1050.1 140.1060.1 140.1052.1 140.1058.1 140.1054.1 140.1056.1 140.1059.1
470001-050EA 86.35 USD
470001-050 470001-040 470001-022 470001-044 470001-030 470001-026 470001-034 470001-046
Preserved Rabbits
Specimens Preserved Specimens Preserved Animal Materials
Standard formalin preserved rabbits, packed in vacuum-sealed bags. One rabbit per bag.
  • Vertebrate anatomy
  • Mammalian anatomy
  • Great alternative to cat dissection
Rabbits are expertly preserved and presented in an extended position for anatomical correctness and ease of study. Our latex injected rabbits are injected with elastic latex with bright, contrasting colors, to ensure ease of dissection. Plastic storage bags and waterproof ID tags are provided free with each rabbit.
Materials Included:
Preserved Rabbit
ID Tag
Specimen Bag
Note: Ward's preserved rabbits are a byproduct of the food industry. Rabbits are not raised specifically for dissection. These animals are "recycled" for educational use.