Genetic Corn Ears
Supplier: Ward's Science
WF-17-6600 17-6500 WF-17-6500
470017-832EA 23.85 USD
470017-832 470017-834 470030-300
Genetic Corn Ears
Specimens Live Specimens Plants - Living Material
Visual examples of genetics at work.
  • F2 ears of varying phenotypes
  • Individually wrapped
Dried and shrink-wrapped ears of corn illustrate naturally occurring Phenotype Ratios of genetic crosses.
3:1 Purple: Yellow: an F2 ear resulting in a P1 having the genotypes R/R and r/r.
3:1 Purple: White: an F2 result of a cross between parental Purple and White. A Single hybrid character with Dominance and Segregation.
9:3:3:1 Purple: White: Smooth: Sweet: an F2 from a parental Purple: Smooth: White: Sweet. Illustrates segregation of two characters.