Ptychodus whipplei (Cretaceous)
Supplier: ADAM MOORE MO
TUCSON0373
470027-456EA 10.5 USD
470027-456
Ptychodus whipplei (Cretaceous)
Characteristic tooth from an early family of fossil skates. Average size: 1/2" in a 1–2" matrix. Texas.