Prepared Phenolphthalein Agar
Supplier: CULTURE MEDIA &SUPPLIES INC SE
880928
470177-378CS 284.95 USD
470177-378
Prepared Phenolphthalein Agar
Media for Microbiology Dehydrated Media for Microbiology
800mL of Phenolphthalein
  • Convenient: no mixing or sterilizing needed
  • Phenolphthalein indicator already included