PCR Based Alu-Human DNA Typing Kit
Supplier: Edvotek
333-A
470112-840EA 129 USD
470112-840
PCR Based Alu-Human DNA Typing Kit
Educational Materials Biology Educational Materials PCR Learning Activities
Using their own hair or cheek cells as sources of DNA, students will isolate human DNA and compare DNA polymorphisms between individuals.
Students use primers for a 300 base pair Alu insertion in chromosome 16 (PV92) to determine their own genotype.
Includes materials for 25 PCR reactions and instructions. A thermal cycler, electrophoresis tank, power supply, variable micropipet and tips, UV transilluminator, balance, and microwave or hot plate are required, but not included.