Paraffin Embedding Table
Supplier: ROCKHARD TOOLS INC SE
E2750.00
470019-026EA 344.25 USD
470019-026
Paraffin Embedding Table
Tissue Embedders
Expand Your Slide Making Skills
  • Quality slide making supplies
  • Create your own slides from your own samples
This compact unit features separated warm and cool sections for melting paraffin, preheating slides, and cooling after embedding, all on the anodized aluminum top. The temperature is adjustable from 45–100°C. 115 VAC, 45 W.