Oersted’s Law Apparatus
Supplier: ELECTRO MECHANICAL INSTRUMENTS
4310
470005-674EA 90.35 USD
470005-674
Oersted’s Law Apparatus
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Ideal for showing the magnetic effects of a current flowing in a wire.
  • Overall dimensions are 30×13×21 cm high
A 15 cm cobalt steel magnetic needle, on an agate bearing, is mounted in a heavy wire frame supported on a stand. The wire frame has three binding posts, two of which are insulated from each other to permit current flow above or below the needle in either direction. Can be operated with any size dry cell battery.
Ordering information: Dry cell battery (not included).