Nitrate-Nitrogen
Supplier: LA MOTTE CHEMICAL PROD CO SE
R-3615 3615-01
470021-172EA 53.55 USD
470021-172 470203-082
Nitrate-Nitrogen
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
Precalibrated test tubes and labware give fast, accurate results on-site, in the classroom, or in the lab.
  • Range: 0 to 1.0 ppm
  • NO3-N cadmium reduction method
  • 40 tests
Reagents, labware, and step-by-step instructions are furnished in a compact, waterproof carrying case.